3-Bromo-4-chlorobenzotrifluoride 97%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2-chloro-5-(trifluoromethyl)benzene, 2-Bromo-1-chloro-4-(trifluoromethyl)benzene, 3-Bromo-4-chloro-α,α,αa-trifluorotoluene

A299160-10G A299160-500G
76772-600EA 9.55 USD
76772-600 76772-602
3-Bromo-4-chlorobenzotrifluoride 97%
3-Bromo-4-chlorobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃BrClF₃
MW: 259.45 g/mol
MDL Number: MFCD00018093
CAS Number: 454-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR