3-Cyano-6-methylchromone 95+%

Supplier: Ambeed
Warning

A299147-1G A299147-250MG A299147-5G
76772-594EA 29 USD
76772-594 76772-596 76772-598
3-Cyano-6-methylchromone 95+%
3-Cyano-6-methylchromone

Precaución: for Research Use Only

Formula: C₁₁H₇NO₂
MDL Number: MFCD00191962
CAS Number: 50743-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR