6-Bromo-4-chloro-7H-pyrrolo[2,3-d]pyrimidine 95%
Supplier: Ambeed
|
Warning
|
A298797-100MG
A298797-1G
A298797-250MG
76772-584EA
51.99
USD
76772-584
76772-586
76772-588
6-Bromo-4-chloro-7H-pyrrolo[2,3-d]pyrimidine 95%
6-Bromo-4-chloro-7H-pyrrolo[2,3-d]pyrimidine
Precaución: for Research Use Only
Formula:
C₆H₃BrClN₃ |
MDL Number:
MFCD08276174 CAS Number: 784150-41-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...