4-Bromo-2,6-difluorobenzonitrile 98%

Supplier: Ambeed
Danger

A298248-10G A298248-1G A298248-250MG A298248-25G A298248-5G
76772-570EA 23 USD
76772-570 76772-572 76772-574 76772-576 76772-578
4-Bromo-2,6-difluorobenzonitrile 98%
4-Bromo-2,6-difluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂BrF₂N
MDL Number: MFCD01310981
CAS Number: 123843-67-4
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR