Methyl-5-aminolevulinate hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Methyl aminolevulinate hydrochloride, δ-Aminolevulinic Acid Methyl Ester Hydrochloride, 3-Aminoacetylpropionic Acid Methyl Ester Hydrochloride, 5-Amino-4-oxovaleric Acid Methyl Ester Hydrochloride, Methyl 5-amino-4-oxopentanoate hydrochloride, 5-Aminolevulinic Acid Methyl Ester Hydrochloride

A298151-10G A298151-1G A298151-250MG A298151-25G A298151-25MG A298151-5G A298151-100G
76772-558EA 49 USD
76772-558 76772-560 76772-562 76772-564 76772-566 76772-568 77877-969
Methyl-5-aminolevulinate hydrochloride 98%
Methyl-5-aminolevulinate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁NO₃
MDL Number: MFCD00012868
CAS Number: 79416-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR