5-Amino-1-(p-tolyl)-1H-pyrazole-4-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-1-(4-methylphenyl)-1H-pyrazole-4-carboxylic acid

A297967-1G A297967-250MG A297967-5G
76772-550EA 48 USD
76772-550 76772-552 76772-554
5-Amino-1-(p-tolyl)-1H-pyrazole-4-carboxylic acid 98%
5-Amino-1-(p-tolyl)-1H-pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁N₃O₂
MW: 217.22 g/mol
MDL Number: MFCD00973937
CAS Number: 14678-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR