5-Chloro-1-methyl-1H-pyrazole-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A297272-10G A297272-1G A297272-250MG A297272-5G
76772-540EA 223.85 USD
76772-540 76772-542 76772-544 76772-546
5-Chloro-1-methyl-1H-pyrazole-4-carboxylic acid 98%
5-Chloro-1-methyl-1H-pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O₂
MDL Number: MFCD02089694
CAS Number: 54367-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR