6-Chloro-2-pyridineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Chloropyridine-2-boronic acid pinacol ester

A296735-1G A296735-250MG A296735-5G A296735-10G A296735-25G
76772-532EA 37 USD
76772-532 76772-534 76772-536 77877-964 77877-965
6-Chloro-2-pyridineboronic acid pinacol ester 98%
6-Chloro-2-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₅BClNO₂
MDL Number: MFCD06798259
CAS Number: 652148-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR