5-Chlorovanillic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-hydroxy-5-methoxybenzoic acid

A296204-25G A296204-5G A296204-1G
76772-518EA 175 USD
76772-518 76772-520 77877-959
5-Chlorovanillic acid 95%
5-Chlorovanillic acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₄
MW: 202.59
MDL Number: MFCD00016531
CAS Number: 62936-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR