6-Formylindolo[3,2-b]carbazole 98%

Supplier: Ambeed
Warning

Synonyms: FICZ, 5,11-Dihydroindolo[3,2-b]carbazole-6-carbaldehyde

A296079-1MG A296079-5MG
76772-514EA 89.12 USD
76772-514 76772-516
6-Formylindolo[3,2-b]carbazole 98%
6-Formylindolo[3,2-b]carbazole

Precaución: for Research Use Only

Formula: C₁₉H₁₂N₂O
MW: 284.32 g/mol
MDL Number: MFCD09879259
CAS Number: 172922-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR