5-Methyl-2-phenyl-1,3-oxazole-4-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-Methyl-2-phenyloxazole-4-carboxylic acid

A294838-1G A294838-250MG A294838-5G
76772-508EA 74.27 USD
76772-508 76772-510 76772-512
5-Methyl-2-phenyl-1,3-oxazole-4-carboxylic acid 97%
5-Methyl-2-phenyl-1,3-oxazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₉NO₃
MW: 203.2 g/mol
MDL Number: MFCD00275475
CAS Number: 18735-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR