4-tert-Butyl-2-chlorophenol 95%

Supplier: Ambeed
Warning

Synonyms: 4-(tert-Butyl)-2-chlorophenol

A294750-1G A294750-25G A294750-5G
76772-502EA 10 USD
76772-502 76772-504 76772-506
4-tert-Butyl-2-chlorophenol 95%
4-tert-Butyl-2-chlorophenol

Precaución: for Research Use Only

Formula: C₁₀H₁₃ClO
MW: 184.66 g/mol
MDL Number: MFCD00019992
CAS Number: 98-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR