5-Chloro-2-nitrophenol 97%

Supplier: Ambeed
Danger

A293668-100G A293668-10G A293668-1G A293668-25G A293668-5G
76772-474EA 149 USD
76772-474 76772-476 76772-478 76772-480 76772-482
5-Chloro-2-nitrophenol 97%
5-Chloro-2-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₄ClNO₃
MDL Number: MFCD09260849
CAS Number: 611-07-4
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR