5-Methoxy-2-tetralone 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,4-Dihydro-5-methoxy-2(1H)-naphthalenone
A292754-100G
A292754-10G
A292754-1G
A292754-25G
A292754-500G
A292754-5G
76772-456EA
300.24
USD
76772-456
76772-458
76772-460
76772-462
76772-464
76772-466
5-Methoxy-2-tetralone 95%
5-Methoxy-2-tetralone
Precaución: for Research Use Only
Formula:
C₁₁H₁₂O₂ MW: 176.22 g/mol |
MDL Number:
MFCD00064958 CAS Number: 32940-15-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...