4-Fluoro-m-toluidine 98%

Supplier: Ambeed
Warning

A291024-25G A291024-500G A291024-100G A291024-10G
76772-450EA 11 USD
76772-450 76772-452 76772-454 77877-949 77877-950
4-Fluoro-m-toluidine 98%
4-Fluoro-m-toluidine

Precaución: for Research Use Only

Formula: C₇H₈FN
MDL Number: MFCD00025294
CAS Number: 452-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR