4-Amino-N,N-diethylaniline 98%

Supplier: Ambeed
Danger

Synonyms: N,N-Diethyl-1,4-phenylenediamine, N,N-Diethyl-p-phenylenediamine, N,N-Diethylbenzene-1,4-diamine, N,N-Diethylbenzene-p-diamine, N-1,N-1-Diethyl-1,4-benzenediamine

A290561-1G A290561-25G A290561-5G
76772-442EA 12.74 USD
76772-442 76772-444 76772-446
4-Amino-N,N-diethylaniline 98%
4-Amino-N,N-diethylaniline

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂
MW: 164.25 g/mol
MDL Number: MFCD00007861
CAS Number: 93-05-0
UN: 2922
ADR: 8,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR