5-Bromo-3-iodo-1H-indole, N-BOC protected 98%

Supplier: Ambeed
Warning

A289054-1G A289054-250MG A289054-25G A289054-5G
76772-434EA 20 USD
76772-434 76772-436 76772-438 76772-440
5-Bromo-3-iodo-1H-indole, N-BOC protected 98%
5-Bromo-3-iodo-1H-indole, N-BOC protected

Precaución: for Research Use Only

Formula: C₁₃H₁₃BrINO₂
MDL Number: MFCD05864761
CAS Number: 850349-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR