2-Bromo-5-(trifluoromethyl)benzonitrile 98%

Supplier: Ambeed
Warning

A288374-1G A288374-5G
76772-424EA 8.49 USD
76772-424 77626-492
2-Bromo-5-(trifluoromethyl)benzonitrile 98%
2-Bromo-5-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₃BrF₃N
MDL Number: MFCD03412187
CAS Number: 1483-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR