5-Nitro-1H-pyrrolo[2,3-b]pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Nitro-7-azaindole

A287439-100MG A287439-1G A287439-250MG A287439-25G A287439-5G
76772-414EA 13.8 USD
76772-414 76772-416 76772-418 76772-420 76772-422
5-Nitro-1H-pyrrolo[2,3-b]pyridine 97%
5-Nitro-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C7H5N3O2
MW: 163.1335 g/mol
MDL Number: MFCD06659682
CAS Number: 101083-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR