5-Bromo-2-chloro-1H-1,3-benzimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-chloro-1H-benzo[d]imidazole

A286219-1G A286219-250MG A286219-5G
76772-400EA 31.83 USD
76772-400 76772-402 76772-404
5-Bromo-2-chloro-1H-1,3-benzimidazole 98%
5-Bromo-2-chloro-1H-1,3-benzimidazole

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MW: 231.49 g/mol
MDL Number: MFCD04128987
CAS Number: 683240-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR