5-(Trimethylsilyl)thiazole 98%

Supplier: Ambeed
Warning

A285198-10G A285198-1G A285198-250MG A285198-5G
76772-370EA 442.4 USD
76772-370 76772-372 76772-374 76772-376
5-(Trimethylsilyl)thiazole 98%
5-(Trimethylsilyl)thiazole

Precaución: for Research Use Only

Formula: C₆H₁₁NSSi
MW: 157.31 g/mol
MDL Number: MFCD00191506
CAS Number: 79265-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR