4-Bromo-5-nitro-1H-imidazole 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-5-nitroimidazole

A283893-250MG A283893-1G A283893-5G
76772-362EA 29.71 USD
76772-362 77593-744 77598-396
4-Bromo-5-nitro-1H-imidazole 97%
4-Bromo-5-nitro-1H-imidazole

Precaución: for Research Use Only

Formula: C₃H₂BrN₃O₂
MW: 191.971 g/mol
MDL Number: MFCD11501602
CAS Number: 6963-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR