4-Bromo-3-fluoroanisole 97%

Supplier: Ambeed
Warning

A283573-100G A283573-10G A283573-1G A283573-25G A283573-500G A283573-5G
76772-344EA 86 USD
76772-344 76772-346 76772-348 76772-350 76772-352 76772-354
4-Bromo-3-fluoroanisole 97%
4-Bromo-3-fluoroanisole

Precaución: for Research Use Only

Formula: C₇H₆BrFO
MDL Number: MFCD01310983
CAS Number: 458-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR