2-Fluoro-6-iodobenzaldehyde 95%

Supplier: Ambeed
Danger

Synonyms: 6-Fluoro-2-Iodobenzaldehyde

A283358-250MG A283358-5G A283358-1G
76772-340EA 24.41 USD
76772-340 76772-342 77626-450
2-Fluoro-6-iodobenzaldehyde 95%
2-Fluoro-6-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄FIO
MW: 250.01 g/mol
MDL Number: MFCD07371506
CAS Number: 146137-72-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR