2-Isopropyl-4-(methylaminomethyl)thiazole 98%

Supplier: Ambeed
Warning

A283288-1G A283288-25G A283288-5G
76772-336EA 10.61 USD
76772-336 76772-338 77571-758
2-Isopropyl-4-(methylaminomethyl)thiazole 98%
2-Isopropyl-4-(methylaminomethyl)thiazole

Precaución: for Research Use Only

Formula: C₈H₁₄N₂S
MW: 170.2752 g/mol
MDL Number: MFCD06797969
CAS Number: 154212-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR