2-Chloro-6-fluorobenzamide 98%

Supplier: Ambeed
Warning

A282879-100G A282879-25G A282879-5G
76772-320EA 106 USD
76772-320 76772-322 76772-324
2-Chloro-6-fluorobenzamide 98%
2-Chloro-6-fluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₅ClFNO
MDL Number: MFCD00055436
CAS Number: 66073-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR