5-Chloro-3-iodo-2-methylaniline 98%

Supplier: Ambeed
Warning

A281771-1G A281771-250MG
76772-316EA 111.4 USD
76772-316 76772-318
5-Chloro-3-iodo-2-methylaniline 98%
5-Chloro-3-iodo-2-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇ClIN
MW: 267.50 g/mol
MDL Number: MFCD08690116
CAS Number: 870606-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR