6,7-Dihydro-5H-pyrrolo[1,2-a]imidazole-2-carboxaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 6,7-Dihydro-2-formyl-5H-pyrrolo[1,2-a]imidazole

A281557-100MG A281557-1G
76772-312EA 203.7 USD
76772-312 76772-314
6,7-Dihydro-5H-pyrrolo[1,2-a]imidazole-2-carboxaldehyde 97%
6,7-Dihydro-5H-pyrrolo[1,2-a]imidazole-2-carboxaldehyde

Precaución: for Research Use Only

Formula: C₇H₈N₂O
MW: 136.151 g/mol
MDL Number: MFCD11035720
CAS Number: 623564-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR