2'-Chloro-5'-fluoroacetophenone 97%

Supplier: Ambeed
Warning

A281450-1G A281450-25G A281450-5G
76772-298EA 13.8 USD
76772-298 76772-300 76772-302
2'-Chloro-5'-fluoroacetophenone 97%
2'-Chloro-5'-fluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆ClFO
MW: 172.59 g/mol
MDL Number: MFCD00042574
CAS Number: 2965-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR