N-Boc-6-Bromohexylamine 97%

Supplier: Ambeed
Danger

Synonyms: tert-Butyl (6-bromohexyl)carbamate, Boc-NH-C6-Br

A279825-1G A279825-250MG A279825-5G A279825-10G A279825-25G A279825-100G
76772-288EA 5.31 USD
76772-288 76772-290 76772-292 76772-294 77568-560 77571-742 77626-442
N-Boc-6-Bromohexylamine 97%
N-Boc-6-Bromohexylamine

Precaución: For research use only.

Formula: C₁₁H₂₂BrNO₂
MDL Number: MFCD06201019
CAS Number: 142356-33-0
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR