5,5-Bis(hydroxymethyl)-2-phenyl-1,3-dioxane 98%

Supplier: Ambeed
Warning

A279718-1G A279718-250MG A279718-5G
76772-274EA 43.5 USD
76772-274 76772-276 76772-278
5,5-Bis(hydroxymethyl)-2-phenyl-1,3-dioxane 98%
5,5-Bis(hydroxymethyl)-2-phenyl-1,3-dioxane

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₄
MDL Number: MFCD00085385
CAS Number: 2425-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR