4-Iodo-3-(trifluoromethyl)aniline 98%

Supplier: Ambeed
Warning

A279532-1G A279532-5G
76772-270EA 17 USD
76772-270 76772-272
4-Iodo-3-(trifluoromethyl)aniline 98%
4-Iodo-3-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₅F₃IN
MDL Number: MFCD04972659
CAS Number: 155403-06-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR