5-Bromo-2-chloro-m-xylene 98%

Supplier: Ambeed
Warning

A277127-100G A277127-25G A277127-5G
76772-250EA 130 USD
76772-250 76772-252 76772-254
5-Bromo-2-chloro-m-xylene 98%
5-Bromo-2-chloro-m-xylene

Precaución: for Research Use Only

Formula: C₈H₈BrCl
MDL Number: MFCD03094622
CAS Number: 206559-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR