5-Norbornen-2-yl acetate (endo- and exo- mixture) 95%

Supplier: Ambeed
Warning

Synonyms: 5-Norbornene-2-yl acetate, Bicyclo[2.2.1]hept-5-en-2-yl Acetate, Acetic Acid Bicyclo[2.2.1]hept-5-en-2-yl Ester, Acetic Acid 5-Norbornen-2-yl Ester

A276242-25G A276242-5G A276242-100G A276242-10G
76772-234EA 5.31 USD
76772-234 76772-236 77571-708 77877-920 77877-921
5-Norbornen-2-yl acetate (endo- and exo- mixture) 95%
5-Norbornen-2-yl acetate (endo- and exo- mixture)

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MDL Number: MFCD00167565
CAS Number: 6143-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR