5-(4-Chlorophenyl)isoxazole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A276119-1G A276119-250MG A276119-5G
76772-228EA 67.9 USD
76772-228 76772-230 76772-232
5-(4-Chlorophenyl)isoxazole-3-carboxylic acid 95%
5-(4-Chlorophenyl)isoxazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆ClNO₃
MW: 223.62 g/mol
MDL Number: MFCD06010034
CAS Number: 33282-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR