Cyclopentanecarbonitrile 98%

Supplier: Ambeed
Danger

A275048-25G A275048-5G
76772-214EA 78.51 USD
76772-214 76772-216
Cyclopentanecarbonitrile 98%
Cyclopentanecarbonitrile

Precaución: for Research Use Only

Formula: C₆H₉N
MW: 95.14 g/mol
MDL Number: MFCD00013741
CAS Number: 4254-02-8
UN: 1992
ADR: 3,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR