2-Fluoro-6-iodotoluene 98%

Supplier: Ambeed
Warning

A272641-100G A272641-1G A272641-25G A272641-5G
76772-154EA 159.14 USD
76772-154 76772-156 76772-158 76772-160
2-Fluoro-6-iodotoluene 98%
2-Fluoro-6-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₆FI
MDL Number: MFCD00142922
CAS Number: 443-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR