3-Amino-4-(isobutylamino)quinoline hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: N4-Isobutylquinoline-3,4-diamine hydrochloride, 3-Amino-4-(isobutylamino)quinoline Hydrochloride

A270511-1G A270511-25G A270511-5G
76772-134EA 9.55 USD
76772-134 76772-136 76772-138
3-Amino-4-(isobutylamino)quinoline hydrochloride 97%
3-Amino-4-(isobutylamino)quinoline hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₁₇N₃
MW: 251.76 g/mol
MDL Number: MFCD08063180
CAS Number: 935521-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR