5-Bromo-1,2-dimethyl-1H-imidazole 97%
Supplier: Ambeed
|
Warning
|
A270090-10G
A270090-1G
A270090-250MG
A270090-5G
A270090-25G
76772-124EA
88
USD
76772-124
76772-126
76772-128
76772-130
77877-904
5-Bromo-1,2-dimethyl-1H-imidazole 97%
5-Bromo-1,2-dimethyl-1H-imidazole
Precaución: for Research Use Only
Formula:
C₅H₇BrN₂ MW: 175.026 g/mol |
MDL Number:
MFCD06659905 CAS Number: 24134-09-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...