6-Fluoro-2-methyl-4-quinolinol 96%

Supplier: Ambeed
Warning

Synonyms: 6-Fluoro-4-hydroxy-2-methylquinoline, 6-Fluoro-2-methylquinolin-4-ol

A267667-1G A267667-250MG A267667-5G
76772-086EA 47.75 USD
76772-086 76772-088 76772-090
6-Fluoro-2-methyl-4-quinolinol 96%
6-Fluoro-2-methyl-4-quinolinol

Precaución: for Research Use Only

Formula: C₁₀H₈FNO
MW: 177.18 g/mol
MDL Number: MFCD00041442
CAS Number: 15912-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR