6-Chloro-5-(2-chloroethyl)oxindole 97%

Supplier: Ambeed
Danger

Synonyms: 6-Chloro-5-(2-chloroethyl)indolin-2-one, Ziprasidone Impurity F, 6-Chloro-5-(2-chloroethyl)-2-indolinone

A267453-1G A267453-250MG A267453-25G A267453-5G
76772-078EA 9.55 USD
76772-078 76772-080 76772-082 76772-084
6-Chloro-5-(2-chloroethyl)oxindole 97%
6-Chloro-5-(2-chloroethyl)oxindole

Precaución: for Research Use Only

Formula: C₁₀H₉Cl₂NO
MW: 230.09 g/mol
MDL Number: MFCD03411598
CAS Number: 118289-55-7
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR