(R)-3-(Boc-amino)piperidine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl N-[(3R)-piperidin-3-yl]carbamate, (R)-3-Boc-aminopiperidine, (R)-tert-Butyl piperidin-3-ylcarbamate
A267248-100G
A267248-10G
A267248-25G
A267248-5G
A267248-500G
76772-056EA
101
USD
76772-056
76772-058
76772-060
76772-062
77598-384
(R)-3-(Boc-amino)piperidine 97%
(R)-3-(Boc-amino)piperidine
Precaución: for Research Use Only
Formula:
C₁₀H₂₀N₂O₂ |
MDL Number:
MFCD03093382 CAS Number: 309956-78-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...