Methyl-2-oxo-2,3-dihydro-1H-1,3-benzimidazole-5-carboxylate 95%

Supplier: Ambeed
Warning

A266432-10G A266432-1G A266432-250MG A266432-25G A266432-5G
76772-016EA 81 USD
76772-016 76772-018 76772-020 76772-022 76772-024
Methyl-2-oxo-2,3-dihydro-1H-1,3-benzimidazole-5-carboxylate 95%
Methyl-2-oxo-2,3-dihydro-1H-1,3-benzimidazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈N₂O₃
MDL Number: MFCD08457979
CAS Number: 106429-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR