1,3-Dimethyl-1H-pyrazole-5-carboxamide 97%

Supplier: Ambeed
Warning

A266038-1G A266038-5G
76772-006EA 26.53 USD
76772-006 76772-008
1,3-Dimethyl-1H-pyrazole-5-carboxamide 97%
1,3-Dimethyl-1H-pyrazole-5-carboxamide

Precaución: for Research Use Only

Formula: C₆H₉N₃O
MW: 139.155 g/mol
MDL Number: MFCD00833259
CAS Number: 136678-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR