6-Benzyl-2-chloro-5,6,7,8-tetrahydro-1,6-naphthyridine 98%

Supplier: Ambeed
Warning

A265491-100MG A265491-1G A265491-250MG
76771-992EA 48.81 USD
76771-992 76771-994 76771-996
6-Benzyl-2-chloro-5,6,7,8-tetrahydro-1,6-naphthyridine 98%
6-Benzyl-2-chloro-5,6,7,8-tetrahydro-1,6-naphthyridine

Precaución: for Research Use Only

Formula: C₁₅H₁₅ClN₂
MDL Number: MFCD09040695
CAS Number: 210539-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR