5-(Trifluoromethyl)pyridin-2-ylamidoxime 95+%

Supplier: Ambeed
Warning

Synonyms: 5-(Trifluoromethyl)pyridine-2-carboxamide oxime

A264995-100MG A264995-1G A264995-250MG
76771-986EA 63 USD
76771-986 76771-988 76771-990
5-(Trifluoromethyl)pyridin-2-ylamidoxime 95+%
5-(Trifluoromethyl)pyridin-2-ylamidoxime

Precaución: for Research Use Only

Formula: C₇H₆F₃N₃O
MDL Number: MFCD00205809
CAS Number: 175277-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR