(S)-5,7-Dihydroxy-2-(4-hydroxyphenyl)-6,8-dimethylchroman-4-one 98%

Supplier: Ambeed
Warning

Synonyms: Farrerol

A264602-100MG A264602-250MG
76771-968EA 129.43 USD
76771-968 76771-970
(S)-5,7-Dihydroxy-2-(4-hydroxyphenyl)-6,8-dimethylchroman-4-one 98%
(S)-5,7-Dihydroxy-2-(4-hydroxyphenyl)-6,8-dimethylchroman-4-one

Precaución: for Research Use Only

Formula: C₁₇H₁₆O₅
MW: 300.31 g/mol
MDL Number: MFCD28347824
CAS Number: 24211-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR