1-Boc-3-iodoindole-5-carbonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 5-Cyano-3-iodo-1H-indole, N-BOC protected

A264107-100G A264107-1G A264107-25G A264107-5G
76771-958EA 1239.14 USD
76771-958 76771-960 76771-962 76771-964
1-Boc-3-iodoindole-5-carbonitrile 98%
1-Boc-3-iodoindole-5-carbonitrile

Precaución: for Research Use Only

Formula: C₁₄H₁₃IN₂O₂
MW: 368.174 g/mol
MDL Number: MFCD05864767
CAS Number: 864685-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR