5-Chlorobenzoxazole-2-thiol 98%

Supplier: Ambeed
Warning

Synonyms: 5-Chlorobenzo[d]oxazole-2(3H)-thione

A263117-25G A263117-5G
76771-930EA 39.26 USD
76771-930 76771-932
5-Chlorobenzoxazole-2-thiol 98%
5-Chlorobenzoxazole-2-thiol

Precaución: for Research Use Only

Formula: C₇H₄ClNOS
MW: 185.63 g/mol
MDL Number: MFCD00175241
CAS Number: 22876-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR