6-Hydroxy-1,4-benzodioxane 97%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dihydrobenzo[b][1,4]dioxin-6-ol

A262705-1G A262705-250MG A262705-5G
76771-914EA 51.99 USD
76771-914 76771-916 76771-918
6-Hydroxy-1,4-benzodioxane 97%
6-Hydroxy-1,4-benzodioxane

Precaución: for Research Use Only

Formula: C₈H₈O₃
MW: 152.15 g/mol
MDL Number: MFCD06656571
CAS Number: 10288-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR